Skip to Content
Merck
All Photos(1)

Key Documents

256404

Sigma-Aldrich

1-Propanol

spectrophotometric grade, ≥99%

Synonym(s):

Propyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2OH
CAS Number:
Molecular Weight:
60.10
Beilstein:
1098242
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

grade

spectrophotometric grade

vapor density

2.1 (vs air)

vapor pressure

10 mmHg ( 147 °C)
14.9 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

700 °F

expl. lim.

13.7 %

impurities

<0.080% water

evapn. residue

<0.0005%

refractive index

n20/D 1.384 (lit.)

bp

97 °C (lit.)
97 °C

mp

−127 °C (lit.)

density

0.804 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 210 nm Amax: 1.00
λ: 225 nm Amax: 0.50
λ: 250 nm Amax: 0.05
λ: 300-400 nm Amax: 0.01

SMILES string

CCCO

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3

InChI key

BDERNNFJNOPAEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anja K K Rahn et al.
Journal of agricultural and food chemistry, 61(37), 8743-8751 (2013-08-24)
Chloropropanol (CP) esters are part of an emerging group of process-induced toxicants that are considered as potential health hazards particularly in palm oil. Mass spectrometry-based methodologies for identification of CP esters in food are critical in overcoming the challenges associated
Dieuwke Sevenster et al.
Science (New York, N.Y.), 339(6121), 830-833 (2013-02-16)
Although reconsolidation opens up new avenues to erase excessive fear memory, subtle boundary conditions put constraints on retrieval-induced plasticity. Reconsolidation may only take place when memory reactivation involves an experience that engages new learning (prediction error). Thus far, it has
Sheba D Bergman et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(33), 10393-10398 (2012-07-13)
A general directed Ru-catalyzed C(sp(3))-H α-alkylation protocol for piperidines (less-reactive substrates than the corresponding five-membered cyclic amines) has been developed. The use of a hindered alcohol (2,4-dimethyl-3-pentanol) as the solvent and catalyst activator, and a catalytic amount of trans-1,2-cyclohexanedicarboxylic acid
Elias N Glaros et al.
The Journal of biological chemistry, 280(26), 24515-24523 (2005-05-14)
Cellular glycosphingolipid (GSL) storage is known to promote cholesterol accumulation. Although physical interactions between GSLs and cholesterol are thought to cause intracellular cholesterol "trapping," it is not known whether cholesterol homeostatic mechanisms are also impaired under these conditions. ApoA-I-mediated cholesterol
Takeshi Morita et al.
The journal of physical chemistry. B, 116(24), 7328-7333 (2012-06-01)
We characterized the effects of ethanol (ET) and dimethyl sulfoxide (DMSO) on H(2)O within a limited H(2)O-rich region by the 1-propanol (1P)-probing methodology developed by us earlier. The results are displayed on a two-dimensional map with twin coordinates: one pertaining

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service