Skip to Content
Merck
All Photos(2)

Key Documents

252883

Sigma-Aldrich

5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine

Dye content 95 %

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€121.00

€121.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€121.00

About This Item

Empirical Formula (Hill Notation):
C48H38N4O4
CAS Number:
Molecular Weight:
734.84
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€121.00


Check Cart for Availability

Request a Bulk Order

form

powder or crystals

Quality Level

composition

Dye content, 95%

λmax

424 nm
653 nm (2nd)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

COc1ccc(cc1)-c2c3ccc(n3)c(-c4ccc(OC)cc4)c5ccc([nH]5)c(-c6ccc(OC)cc6)c7ccc(n7)c(-c8ccc(OC)cc8)c9ccc2[nH]9

InChI

1S/C48H38N4O4/c1-53-33-13-5-29(6-14-33)45-37-21-23-39(49-37)46(30-7-15-34(54-2)16-8-30)41-25-27-43(51-41)48(32-11-19-36(56-4)20-12-32)44-28-26-42(52-44)47(40-24-22-38(45)50-40)31-9-17-35(55-3)18-10-31/h5-28,49,52H,1-4H3/b45-37-,45-38-,46-39-,46-41-,47-40-,47-42-,48-43-,48-44-

InChI key

PJOJZHHAECOAFH-PJEPRTEXSA-N

General description

5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine is an achiral porphyrin derivative. It forms chiral supramolecular assemblies via spontaneous symmetry breaking at the air or water interface.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Belén Ballatore et al.
European journal of medicinal chemistry, 83, 685-694 (2014-07-11)
A covalently linked porphyrin-fullerene C60 dyad 5 was synthesized by 1,3-dipolar cycloaddition using 5-(4-formylphenyl)-10,15,20-tris[3-(N-ethylcarbazoyl)]porphyrin, N-methylglycine and fullerene C60. Methylation of 5 was used to obtain a cationic dyad 6. Spectroscopic properties were compared in toluene, N,N-dimethylformamide (DMF) and toluene/sodium bis(2-ethylhexyl)sulfosuccinate
Maximiliano L Agazzi et al.
Organic & biomolecular chemistry, 18(7), 1449-1461 (2020-02-07)
Chromophore-fullerene C60 hybrids possess interesting properties that enable them to act as heavy atom-free photosensitizers and reactive oxygen species (ROS) producers. Here, two new diketopyrrolopyrrole-C60 conjugates were efficiently synthesized and characterized. The conjugates show broadband absorption in the visible spectral
Chirality amplification of porphyrin assemblies exclusively constructed from achiral porphyrin derivatives.
Chen P, et al.
ChemPhysChem, 7(11), 2419-2423 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service