Skip to Content
Merck
All Photos(1)

Key Documents

20860

Supelco

Cadion

for spectrophotometric det. of Cd, Ni, ≥98.0%

Synonym(s):

1-(4-Nitrophenyl)-3-(4-phenylazophenyl)triazene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14N6O2
CAS Number:
Molecular Weight:
346.34
Beilstein:
935340
EC Number:
MDL number:
UNSPSC Code:
41115717
PubChem Substance ID:

Quality Level

Assay

≥98.0% (TLC)
≥98.0%

form

powder

quality

for spectrophotometric det. of Cd, Ni

technique(s)

UV/Vis spectroscopy: suitable

SMILES string

[O-][N+](=O)c1ccc(cc1)\N=N\Nc2ccc(cc2)\N=N\c3ccccc3

InChI

1S/C18H14N6O2/c25-24(26)18-12-10-17(11-13-18)22-23-21-16-8-6-15(7-9-16)20-19-14-4-2-1-3-5-14/h1-13H,(H,21,22)/b20-19+

InChI key

SMMIDVLUFMPWFN-FMQUCBEESA-N

Application

Cadion along with p-nitrobenzenediazoaminobenzene-p-azobenzene and Triton X-100 may be used in determination of Cadmium by dual-wavelength spectrophotometry.

Other Notes

Reagent for the determination of Cadmium(II)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Watanabe et al.
Talanta, 26(10), 959-961 (1979-10-01)
Cadmium has been determined by dual-wavelength spectrophotometry with cadion, p-nitrobenzenediazoaminobenzene-p-azobenzene, and a non-ionic surfactant, Triton X-100. Cadion and its cadmium chelate are dissolved in a micellar solution of the surfactant. The absorbance difference at the wavelength of maximum absorption of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service