Skip to Content
Merck
All Photos(2)

Key Documents

201723

Sigma-Aldrich

Plasmocorinth B

Dye content 60 %

Synonym(s):

3-(5-Chloro-2-hydroxyphenylazo)-4,5-dihydroxynaphthalene-2,7-disulfonic acid disodium salt, Calcium fast purple, Michrome No. 452, Mordant Blue 13

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H9ClN2Na2O9S2
CAS Number:
Molecular Weight:
518.81
Colour Index Number:
16680
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 60%

λmax

527 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Oc1ccc(Cl)cc1\N=N\c2c(O)c3c(O)cc(cc3cc2S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C16H11ClN2O9S2.2Na/c17-8-1-2-11(20)10(5-8)18-19-15-13(30(26,27)28)4-7-3-9(29(23,24)25)6-12(21)14(7)16(15)22;;/h1-6,20-22H,(H,23,24,25)(H,26,27,28);;/q;2*+1/p-2/b19-18+;;

InChI key

LNXMADNIUWFTPP-BUFQOAPZSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Heinonen et al.
Neuroscience letters, 72(1), 105-110 (1986-12-03)
Cytoplasmic, free Mg2+ was measured spectrophotometrically using an intrasynaptosomally entrapped Mg2+-indicator, Eriochrome blue (EB). Addition of the ionophore A23187 or disruption of the synaptosomal plasma membrane with digitonin caused an increase in absorbance of entrapped EB with a maximum at
E Heinonen et al.
Biochimica et biophysica acta, 898(3), 331-337 (1987-04-23)
The free Mg2+ concentration within synaptosomes has been measured with an entrapped Mg2+ indicator, eriochrome blue. Ionophores gramicidin and A23187 slowly increased the absorbance of the entrapped dye. Calibration of the dye response in a Na+-based medium gave a value
Measurement of ionized magnesium in biological fluids.
E Casillas et al.
Analytical biochemistry, 116(2), 319-324 (1981-09-15)
A Scarpa et al.
Federation proceedings, 40(12), 2646-2652 (1981-10-01)
A nondestructive technique is described for measuring intracellular free Mg2+ concentrations. It is based on the microinjection of a Mg2+ metallochromic indicator in the cytosol of a large single cell and measuring its differential absorbance changes with sensitive multiple wavelength
B Rydqvist et al.
Acta physiologica Scandinavica, 127(4), 499-506 (1986-08-01)
Intracellular free Mg2+ (Mgf) in darkness and during illumination was measured in the Balanus photoreceptor using the Mg2+ sensitive dye Eriochrome Blue SE (EB). The EB was pressure injected in the cell and Mgf determined from measurements of absorption changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service