Skip to Content
Merck
All Photos(3)

Key Documents

192457

Sigma-Aldrich

Cyclohexanone 2,4-dinitrophenylhydrazone

≥99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€144.00

€144.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€144.00

About This Item

Linear Formula:
C6H10[=NNHC6H3(NO2)2]
CAS Number:
Molecular Weight:
278.26
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€144.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

≥99%

form

powder or crystals

mp

159-160 °C (lit.)

solubility

chloroform: 25 mg/mL, clear, orange

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[O-][N+](=O)c1ccc(N\N=C2\CCCCC2)c(c1)[N+]([O-])=O

InChI

1S/C12H14N4O4/c17-15(18)10-6-7-11(12(8-10)16(19)20)14-13-9-4-2-1-3-5-9/h6-8,14H,1-5H2

InChI key

QLWXZRVOHCYKKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful standard for elemental analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service