Skip to Content
Merck
All Photos(1)

Key Documents

191248

Sigma-Aldrich

3,3′-Dimethoxybenzidine dihydrochloride

technical grade

Synonym(s):

o-Dianisidine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H3(OCH3)NH2]2 · 2HCl
CAS Number:
Molecular Weight:
317.21
Beilstein:
3917996
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

technical grade

Quality Level

form

powder

mp

268 °C (dec.) (lit.)

solubility

water: 25 mg/mL, clear to very slightly hazy

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl.Cl.COc1cc(ccc1N)-c2ccc(N)c(OC)c2

InChI

1S/C14H16N2O2.2ClH/c1-17-13-7-9(3-5-11(13)15)10-4-6-12(16)14(8-10)18-2;;/h3-8H,15-16H2,1-2H3;2*1H

InChI key

UXTIAFYTYOEQHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuro Suzuki et al.
Phytochemistry, 67(3), 219-224 (2006-01-13)
A peroxidase (POX)-containing fraction was purified from buckwheat seed. The POX consisted of two isozymes, POX I and POX II, that were purified 6.6- and 67.4-fold, respectively. Their molecular weights were estimated to be 46.1 kDa (POX I) and 58.1
Rémy Ricoux et al.
Bioconjugate chemistry, 19(4), 899-910 (2008-03-08)
To develop artificial hemoproteins that could lead to new selective oxidation biocatalysts, a strategy based on the insertion of various iron-porphyrin cofactors into Xylanase A (Xln10A) was chosen. This protein has a globally positive charge and a wide enough active
Ozcan Erel
Clinical biochemistry, 38(12), 1103-1111 (2005-10-11)
To develop a new, colorimetric and automated method for measuring total oxidation status (TOS). The assay is based on the oxidation of ferrous ion to ferric ion in the presence of various oxidant species in acidic medium and the measurement
Wei Shen et al.
Biosensors & bioelectronics, 44, 171-176 (2013-02-22)
Herein we report a label-free microRNA (miRNA) biosensor in which the formation of a thin insulating film is used to amplify the analytical signal. Briefly, the biosensor is made of an oligonucleotide-coated gold electrode. After hybridizing with a target miRNA
Uta Merle et al.
Journal of hepatology, 51(5), 925-930 (2009-09-02)
A low serum ceruloplasmin concentration is considered diagnostic for Wilson's disease. We aimed to evaluate an enzymatic test for ceruloplasmin oxidase activity and to compare it with the routinely used immunological ceruloplasmin measurement. Serum ceruloplasmin was measured enzymatically with o-dianisidine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service