Skip to Content
Merck
All Photos(1)

Key Documents

18502

Millipore

Oxidase Reagent according to Gordon-McLeod

suitable for microbiology

Synonym(s):

Gordon-McLeod Reagent

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€62.00

€62.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
100 ML
€62.00

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

€62.00


Check Cart for Availability

Request a Bulk OrderRequest more information

Agency

according to ISO 9308-1:2014

Quality Level

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

composition

N,N-dimethyl-p-phenylenediamine hydrochloride, 0.15 g
water, 10 mL

packaging

pkg of 100 mL

storage condition

(Tightly closed)

technique(s)

microbe id | specific enzyme detection: suitable

antibiotic activity spectrum

Gram-negative bacteria

application(s)

clinical testing
environmental
food and beverages
pharmaceutical

microbiology

suitability

Plesiomonas spp.
Aeromonas spp.
Neisseria spp.
Pseudomonas spp.
bacteria

General description

Oxidase Reagent, according to Gordon-McLeod, is a commonly used laboratory reagent in microbiology to detect bacteria that produce cytochrome c oxidase. The reagent is designed to identify this enzyme′s presence, which plays a critical role in the electron transport chain and indicates respiratory metabolism in bacteria. The Gordon-McLeod reagent contains N,N,N′,N′-tetramethyl-p-phenylenediamine dihydrochloride, as the active component in the reagent. When an organism produces cytochrome c oxidase, the reagent undergoes a color change reaction, turning from colorless to blue or purple. This color change is a positive result, indicating the presence of cytochrome c oxidase activity in the tested organism.

Application

Oxidase Reagent, according to Gordon-McLeod, is a commonly used laboratory technique in industrial microbiology to differentiate between bacterial strains based on their ability to produce cytochrome c oxidase. The Gaby-Hadley A reagent provides a simple and efficient way to identify oxidase-positive bacteria, such as Pseudomonas, Vibrio, and some Neisseria species, while producing negative results for organisms that lack cytochrome c oxidase activity. in the food, beverage, and pharmaceutical industries.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Vibrios are motile, curved or comma-shaped bacilli and have a single polar flagella with sheet proteins. They are often found in open water, freshwater and saltwater.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

Specific agars and broths support detection and cultivation of Campylobacter, requiring complex media with essential supplements.

Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.

See All

Protocols

General guidance for the detection and enumeration of Enterobacteriaceae in food according to EN-ISO 8523:1991 and EN-ISO 4832:1991, respectively.

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service