Skip to Content
Merck
All Photos(1)

Key Documents

183350

Supelco

Poly(ethyl methacrylate)

analytical standard, average Mw 340,000 (Typical), average Mn 126,000 (Typical)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)(CO2C2H5)]n
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

form

crystalline

mol wt

average Mn 126,000 (Typical)
average Mw 340,000 (Typical)

technique(s)

gel permeation chromatography (GPC): suitable

format

neat

SMILES string

CCOC(=O)C(C)=C

InChI

1S/C6H10O2/c1-4-8-6(7)5(2)3/h2,4H2,1,3H3

InChI key

SUPCQIBBMFXVTL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(ethyl methacrylate) was used in combination with PVC in preparing new phase-separated polymer electrolyte. It was used to understand the polymer-solvent, polymer-surfactant and solvent-surfactant interaction effects and the intercalation process in nanocomposite preparation method.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service