Skip to Content
Merck
All Photos(1)

Key Documents

18248

Sigma-Aldrich

Protein Disulfide Isomerase from bovine liver

≥150 U/mg protein (protein ca. 40%)

Synonym(s):

PDI

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

form

powder
powder with small lumps

specific activity

≥150 U/mg protein (protein ca. 40%)

mol wt

107 kDa by gel filtration
57 kDa by SDS-PAGE

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C7H5Cl2NO5S/c8-4-2-5(9)6(16(13,14)15-10)1-3(4)7(11)12/h1-2H,10H2,(H,11,12)

InChI key

DHUYKLYJBKXDBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Physical properties

Protein Disulfide Isomerase (PDI) from bovine liver is a homodimer with a molecular weight of 107 kDa (gel filtration) and the molecular weight of the monomer has been reported at 57 kDA (SDS-PAGE). The enzyme is a glycoprotein with 12% total carbohydrate content, composed of 4.6% mannose, 2.5% galactose, 1.4% NANA, and 3.5% 2-acetamido-2-deoxyglucose.

Unit Definition

1 unit corresponds to the amount of enzyme, which, in a glutathione-dithiothreitol-system, causes a change of absorbance of 0.001 per minute at pH 4.0 and 25°C.

Other Notes

Enzyme that accelerates the exchange reaction between disulfide bridges in proteins

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pryank Patel et al.
Journal of molecular biology, 425(5), 929-943 (2013-01-01)
Anterior gradient 2 (AGR2) is a normal endoplasmic reticulum protein that has two important abnormal functions, amphibian limb regeneration and human cancer metastasis promotion. These normal intracellular and abnormal extracellular roles can be attributed to the multidomain structure of AGR2.
Harshavardhan Kenche et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(3), 965-977 (2012-11-22)
The endoplasmic reticulum (ER) stress response (ERSR) and associated protein aggregation, is under investigation for its role in human diseases, including chronic obstructive pulmonary disease (COPD) where cigarette smoking (CS) is a risk factor for disease development. Our hypothesis states
James J Galligan et al.
Human genomics, 6, 6-6 (2012-12-19)
Enzyme-mediated disulfide bond formation is a highly conserved process affecting over one-third of all eukaryotic proteins. The enzymes primarily responsible for facilitating thiol-disulfide exchange are members of an expanding family of proteins known as protein disulfide isomerases (PDIs). These proteins
Zegbeh Z Kpadeh et al.
Journal of bacteriology, 195(8), 1825-1833 (2013-02-26)
The extracytoplasmic assembly of the Dot/Icm type IVb secretion system (T4SS) of Legionella pneumophila is dependent on correct disulfide bond (DSB) formation catalyzed by a novel and essential disulfide bond oxidoreductase DsbA2 and not by DsbA1, a second nonessential DSB
Florian Langer et al.
Blood, 121(12), 2324-2335 (2013-01-15)
Lymphocyte depletion with antithymocyte globulin (ATG) can be complicated by systemic coagulation activation. We found that ATG activated tissue factor procoagulant activity (TF PCA) on monocytic cells more potently than other stimuli that decrypt TF, including cell disruption, TF pathway

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service