Skip to Content
Merck
All Photos(1)

Key Documents

181412

Supelco

Poly(butyl acrylate) solution

25 wt. % in toluene, analytical standard, average Mw 60,000 (Typical), average Mn 20,000 (Typical)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C7H12O2)n
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

1.2 (37 °C, vs air)

autoignition temp.

997 °F

mol wt

average Mn 20,000 (Typical)
average Mw 60,000 (Typical)

composition

solids, 25-30 wt. %

concentration

25 wt. % in toluene

technique(s)

gel permeation chromatography (GPC): suitable

refractive index

n20/D 1.466 (lit.)

transition temp

Tg -49 °C

format

single component solution

SMILES string

CCCCOC(=O)C=C

InChI

1S/C7H12O2/c1-3-5-6-9-7(8)4-2/h4H,2-3,5-6H2,1H3

InChI key

CQEYYJKEWSMYFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Unit weight does not include weight of solvent.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service