Skip to Content
Merck
All Photos(1)

Key Documents

18018

Supelco

Allyl cyclohexanepropionate

analytical standard

Synonym(s):

2-Propenyl 3-cyclohexylpropanoate, Allyl 3-cyclohexylpropionate, Cyclohexanepropanoic acid 2-propen-1-yl ester, Cyclohexanepropionic acid allyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH2CH2CO2CH2CH=CH2
CAS Number:
Molecular Weight:
196.29
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.46 (lit.)

bp

91 °C/1 mmHg (lit.)

density

0.948 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

C=CCOC(=O)CCC1CCCCC1

InChI

1S/C12H20O2/c1-2-10-14-12(13)9-8-11-6-4-3-5-7-11/h2,11H,1,3-10H2

InChI key

TWXUTZNBHUWMKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service