Skip to Content
Merck
All Photos(1)

Documents

16016

Sigma-Aldrich

Phenol

puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)

Synonym(s):

Hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
Beilstein:
969616
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21

Agency

complies with USP (identity)

Quality Level

vapor density

3.24 (vs air)

vapor pressure

0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

grade

puriss.

Assay

≥99.5% (GC)
99.0-100.5% (calc. on H2O free substance)

form

crystalline (detached)

autoignition temp.

1319 °F

quality

meets analytical specification of Ph. Eur., BP, USP

expl. lim.

8.6 %

impurities

acidic reac. Impurities, in accordance
organic volatile impurities, in accordance (GC)
residual solvents, complies
≤0.05% non-volatile matter
≤0.3% water (Karl Fischer)

bp

182 °C (lit.)

mp

40-42 °C (lit.)

transition temp

solidification point ≥40 °C

density

1.071 g/mL at 25 °C (lit.)

suitability

in accordance for appearance of solution
Citrobacter spp.
Escherichia coli
coliforms

application(s)

food and beverages
water monitoring

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenol is an aromatic alcohol. Various industrial processes for the synthesis of phenol have been proposed.

Application

Phenol may be employed for the isolation of deoxyribonucleic acid from Bacillus subtilis. It may be used in the Berthelot color reaction for the quantitative estimation of ammonia.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PREPARATION OF TRANSFORMING DEOXYRIBONUCLEIC ACID BY PHENOL TREATMENT.
H SAITO et al.
Biochimica et biophysica acta, 72, 619-629 (1963-08-20)
Tyman JHP.
Synthetic and Natural Phenols., 1-1 (1996)
Phenol-hypochlorite reaction for determination of ammonia.
Weatherburn M W.
Analytical Chemistry, 39(8), 971-974 (1967)
Thitiporn Anunthawan et al.
Biochimica et biophysica acta, 1848(6), 1352-1358 (2015-03-15)
We investigated the mechanisms of two tryptophan-rich antibacterial peptides (KT2 and RT2) obtained in a previous optimization screen for increased killing of both Gram-negative and Gram-positive bacteria pathogens. At their minimal inhibitory concentrations (MICs), these peptides completely killed cells of
Rita A Busuttil et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(10), 2761-2772 (2014-03-25)
Gene-expression profiling has revolutionized the way we think about cancer and confers the ability to observe the synchronous expression of thousands of genes. The use of putative genome-level expression profiles has allowed biologists to observe the complex interactions of genes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service