Skip to Content
Merck
All Photos(1)

Key Documents

15997

Millipore

Brewer Agar

suitable for microbiology

Synonym(s):

Anerobic Agar according to Brewer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

form

solid

shelf life

limited shelf life, expiry date on the label

composition

agar, 12.6 g/L
casein peptone, 10 g/L
L-cystine, 0.4 g/L
D(+)-glucose, 10 g/L
methylene blue, 0.002 g/L
sodium chloride, 5 g/L
sodium formaldehyde sulfoxylate, 1 g/L
sodium thioglycolate, 2 g/L
soy peptone, 5 g/L
yeast extract, 5 g/L

final pH

7.2±0.2 (25 °C)

application(s)

environmental
food and beverages

microbiology

suitability

anaerobic bacteria
nonselective for Bacteroides spp.
nonselective for Clostridium spp.
nonselective for Escherichia coli
nonselective for Staphylococcus spp.
nonselective for coliforms

Application

For the culture of Clostridium species and other anaerobic microorganisms in surface culture according to Brewer.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A NEW PETRI DISH COVER AND TECHNIQUE FOR USE IN THE CULTIVATION OF ANAEROBES AND MICROAEROPHILES.
J H Brewer
Science (New York, N.Y.), 95(2475), 587-587 (1942-06-05)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service