Skip to Content
Merck
All Photos(3)

Key Documents

15971

Supelco

N-Vanillylnonanamide

analytical standard

Synonym(s):

Nonivamide, Capsaicin synthetic, N-(4-Hydroxy-3-methoxybenzyl)nonanamide, N-Vanillylpelargonamide, Nonanoic acid vanillylamide, Nonylvanylamide, Pelargonic acid vanillylamide, Pseudocapsaicin

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€68.80

€68.80


Estimated to ship on17 April 2025


Request a Bulk Order

Select a Size

Change View
50 MG
€68.80

About This Item

Empirical Formula (Hill Notation):
C17H27NO3
CAS Number:
Molecular Weight:
293.40
Beilstein:
2144300
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€68.80


Estimated to ship on17 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCC(=O)NCc1ccc(O)c(OC)c1

InChI

1S/C17H27NO3/c1-3-4-5-6-7-8-9-17(20)18-13-14-10-11-15(19)16(12-14)21-2/h10-12,19H,3-9,13H2,1-2H3,(H,18,20)

InChI key

RGOVYLWUIBMPGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Vanillylnonanamide has antifouling properties. It acts as an nicotinamide adenine dinucleotide phosphate (NADPH) inhibitor.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katalin Horváth et al.
Neuropeptides, 48(6), 371-378 (2014-12-03)
The aim of the present study was to evaluate the therapeutic potential of local capsaicinoid (EMSPOMA(®) cream) treatment on chronic low back pain in patients with degenerative spine diseases and to investigate the possible mechanism of action of the therapy.
Ina Callebaut et al.
American journal of rhinology & allergy, 28(4), 290-296 (2014-09-10)
The naso-ocular interaction in allergic rhinoconjunctivitis is well recognized from epidemiological, clinical, and experimental observations. The precise mechanisms remain incompletely understood. A new mouse model of allergic rhinoconjunctivitis was used to investigate the contribution of mast cells and trigeminal ganglia
Josef Yayan
Medicine, 93(27), e199-e199 (2014-12-17)
Current uses of culture-independent tools in previous studies have shown a significant relationship between microbiota and asthma. Although these studies are relatively new, there is also evidence of the possibility of new therapeutic strategies for the treatment or prevention of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service