Skip to Content
Merck
All Photos(1)

Key Documents

154709

Sigma-Aldrich

Carbon disulfide

spectrophotometric grade, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14
Beilstein:
1098293
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
grade:
spectrophotometric grade
Assay:
≥99%
technique(s):
UV/Vis spectroscopy: suitable
bp:
46 °C (lit.)
vapor pressure:
5.73 psi ( 20 °C)
Pricing and availability is not currently available.

grade

spectrophotometric grade

vapor density

2.67 (vs air)

vapor pressure

5.73 psi ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

212 °F

expl. lim.

50 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.03% water

evapn. residue

<0.0005%

color

APHA: ≤10

refractive index

n20/D 1.627 (lit.)

bp

46 °C (lit.)

mp

−112-−111 °C (lit.)

density

1.266 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 386 nm Amax: 1.0
λ: 388 nm Amax: 0.50
λ: 394 nm Amax: 0.25
λ: 403 nm Amax: 0.10
λ: 410 nm Amax: 0.05
λ: 500-750 nm Amax: 0.01

SMILES string

S=C=S

InChI

1S/CS2/c2-1-3

InChI key

QGJOPFRUJISHPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

Target Organs

Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C60: a new form of carbon.
Kratschmer W, et al.
Nature, 347(6291), 354-358 (1990)
Margherita Maiuri et al.
Physical chemistry chemical physics : PCCP, 14(18), 6312-6319 (2012-02-15)
In carotenoids internal conversion between the allowed (S(2)) and forbidden (S(1)) excited states occurs on a sub-picosecond timescale; the involvement of an intermediate excited state(s) (S(x)) mediating the process is controversial. Here we use high time resolution (sub-20 fs) broadband
[11C]carbon disulfide: a versatile reagent for PET radiolabelling.
Philip W Miller et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(2), 433-436 (2011-12-14)
Marjan J Smeulders et al.
Nature, 478(7369), 412-416 (2011-10-21)
Extremophilic organisms require specialized enzymes for their exotic metabolisms. Acid-loving thermophilic Archaea that live in the mudpots of volcanic solfataras obtain their energy from reduced sulphur compounds such as hydrogen sulphide (H(2)S) and carbon disulphide (CS(2)). The oxidation of these
A critical review of the literature on carbon disulfide toxicity.
R O Beauchamp et al.
Critical reviews in toxicology, 11(3), 169-278 (1983-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service