All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22
EC Number:
MDL number:
UNSPSC Code:
12141751
PubChem Substance ID:
Recommended Products
Assay
≥97.0% Zr + Hf basis
form
powder
reaction suitability
reagent type: catalyst
core: zirconium
resistivity
40 μΩ-cm, 20°C
impurities
≤3% hafnium (Hf)
bp
4377 °C (lit.)
mp
1852 °C (lit.)
density
6.5 g/mL at 25 °C (lit.)
SMILES string
[Zr]
InChI
1S/Zr
InChI key
QCWXUUIWCKQGHC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
bottled under inert gas
Other Notes
A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Pyr. Sol. 1 - Water-react 1
Supplementary Hazards
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Thijs H Oude Munnink et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(6), 929-935 (2013-04-30)
Placental growth factor (PlGF) is a member of the proangiogenic vascular endothelial growth factor family, which is upregulated in many tumors. RO5323441, a humanized monoclonal antibody against PlGF, showed antitumor activity in human tumor xenografts. We therefore aimed to radiolabel
The enigma of the bilayered zirconia restoration.
Bernard E Keough et al.
Dentistry today, 32(5), 68-68 (2013-06-01)
An ideal full-arch tooth replacement option: CAD/CAM zirconia screw-retained implant bridge.
Michael Tischler et al.
Dentistry today, 32(5), 98-102 (2013-06-01)
B Kuter et al.
European journal of paediatric dentistry : official journal of European Academy of Paediatric Dentistry, 14(2), 90-94 (2013-06-14)
The objective of this study was to assess the effect of heat application on the mechanical properties of glass ionomer cements. experimental design. The effect of heat on glass ionomer cements during their setting was evaluated by measuring compressive strength
Removing all-ceramic restorations with lasers.
Jeffrey P Cranska
Dentistry today, 32(6), 101-102 (2013-06-28)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service