Skip to Content
Merck
All Photos(1)

Key Documents

14132

Supelco

Isopentylamine

analytical standard

Synonym(s):

1-Amino-3-methylbutane, Isoamylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2NH2
CAS Number:
Molecular Weight:
87.16
Beilstein:
1209230
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

color

colorless

refractive index

n20/D 1.407-1.409
n20/D 1.408 (lit.)

bp

95-97 °C (lit.)

mp

-83 °C

density

0.751 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(C)CCN

InChI

1S/C5H13N/c1-5(2)3-4-6/h5H,3-4,6H2,1-2H3

InChI key

BMFVGAAISNGQNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Notou et al.
Journal of chromatography. A, 1356, 272-276 (2014-07-06)
A novel zone-fluidics/high pressure liquid chromatographic (ZF-HPLC) method is described for the simultaneous determination of six biogenic monoamines, in the presence of hexylamine as internal standard. Automated on-line derivatization of the analytes with naphthalene-2,3-dicarboxaldehyde/cyanide ions was performed using the ZF

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service