Skip to Content
Merck
All Photos(1)

Key Documents

12257

Sigma-Aldrich

Potassium dichromate

technical, ≥98%

Synonym(s):

Potassium bichromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2Cr2O7
CAS Number:
Molecular Weight:
294.18
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

technical

Assay

≥98%

reaction suitability

reagent type: oxidant

mp

398 °C (lit.)

anion traces

chloride (Cl-): ≤200 mg/kg
sulfate (SO42-): ≤500 mg/kg

SMILES string

[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

InChI

1S/2Cr.2K.7O/q;;2*+1;;;;;;2*-1

InChI key

KMUONIBRACKNSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shafaqat Ali et al.
Environmental toxicology and chemistry, 32(10), 2234-2239 (2013-06-19)
A hydroponic experiment was carried out to examine the effect of hydrogen sulfide (H2 S) in alleviating chromium (Cr) stress in barley. A 2-factorial design with 6 replications was selected, including 3 levels of NaHS (0 μM, 100 μM, and
Lubna Shakir et al.
Environmental toxicology and pharmacology, 34(2), 180-191 (2012-04-24)
The problem of water pollution acquires greater relevance in the context of a developing agrarian economy like Pakistan. Even though, the leather industry is a leading economic sector in Pakistan, there is an increasing environmental concern regarding tanneries because they
Ashish K Mishra et al.
Environmental toxicology, 27(7), 415-422 (2012-06-19)
Exposure to sublethal hexavalent chromium (as 2 and 4 mg L⁻¹ potassium dichromate for 1 and 2 months) during late preparatory to mid prespawning phase of annual reproductive cycle severely affected the pituitary-ovarian axis of a teleost Channa punctatus. Gonadosomatic
Bour-Jr Wang et al.
Journal of the Formosan Medical Association = Taiwan yi zhi, 110(12), 775-779 (2012-01-18)
Occupational dermatitis among cement workers is a major occupational health concern. The two most important occupational hazards for cement workers are irritant and allergic cement contact dermatitis. The objective of this study was to investigate the severity of occupational cement
Fu-Jun Liu et al.
Toxicological sciences : an official journal of the Society of Toxicology, 110(2), 307-318 (2009-06-03)
Werner syndrome is a premature aging disorder characterized by cancer predisposition that is caused by loss of the Werner syndrome protein (WRN) helicase/exonuclease DNA repair protein. Hexavalent chromium is an environmental carcinogen and genotoxicant that is associated with respiratory cancers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service