Skip to Content
Merck
All Photos(1)

Documents

11577

Millipore

Peptone Test Kit with Plant Peptones

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | growth & motility: suitable

application(s)

clinical testing
environmental
food and beverages
pharmaceutical

microbiology

suitability

bacteria
molds
yeasts

General description

Kit contains 20 peptones (10 g each)
18332 Peptone (vegetable)
51841 Peptone (vegetable) acid hydrolysate
19942 Peptone (vegetable), no 1
61854 Peptone (vegetable), no 2
92976 Peptone special (vegetable)
29185 Proteose Peptone (vegetable)
16922 Tryptone (vegetable)
12331 Tryptose (vegetable)
05138 Vegetable Extract
04316 Vegetable Extract no 1
49869 Vegetable Extract no 2
07436 Vegetable hydrolysate no 2
67381 Vegetable Infusion powder
95757 Vegetable Special Infusion powder
83059 Peptone from potatoes
93492 Peptone from wheat
90765 Peptone from soybean, enzymatic digest
70178 Peptone from soybean, enzymatic digest
S1674 Soy protein acid hydrolysate
87972 Peptone enzymatic digest from soybean
Kit contains 20 peptones, extracts and hydrolysates from plant sources. This kit helps to evaluate the best growth properties of microganisms. With a peptide source specifically fitted to a microorganism, it is possible to increase the yield and improve the reproducibility of results.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manal Moussa et al.
BMC biotechnology, 12, 96-96 (2012-12-21)
Currently, the two most commonly used fibrinolytic agents in thrombolytic therapy are recombinant tissue plasminogen activator (rt-PA) and streptokinase (SK). Whereas SK has the advantage of substantially lower costs when compared to other agents, it is less effective than either

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service