Skip to Content
Merck
All Photos(1)

Key Documents

11107

Supelco

α-Asarone

analytical standard

Synonym(s):

trans-1,2,4-Trimethoxy-5-(1-propenyl)benzene, trans-1-Propenyl-2,4,5-trimethoxybenzene, trans-Asarone, Isoasaron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CH=CHCH3
CAS Number:
Molecular Weight:
208.25
Beilstein:
1910606
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

296 °C (lit.)

mp

57-61 °C (lit.)
57-61 °C

application(s)

food and beverages

format

neat

SMILES string

COc1cc(OC)c(\C=C\C)cc1OC

InChI

1S/C12H16O3/c1-5-6-9-7-11(14-3)12(15-4)8-10(9)13-2/h5-8H,1-4H3/b6-5+

InChI key

RKFAZBXYICVSKP-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Asarones are toxic morphogenetic agents. It is also known for its chemosterlant and oviposition stimulant properties. Naturally it can be extracted from Acorus calamus. α-Asarone is considered as growth inhibitor. This isomer acts as antifeedant with less toxicity.

Application

It was used as standard in synthesis of five isomers of α-Asarone to compare the better deterrent using HPLC analysis.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feeding-deterrent activity of a-asarone isomers against some stored Coleoptera.
Poplawski, Janusz, et al.
Pest Management Science, 56.6, 560-564 (2000)
Zhenwei Yu et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 36(21), 2967-2970 (2012-02-09)
To investigate the relationship of properties and drug release rate of hot melt pressure sensitive adhesive (HMPSA), and to provide a recommendation of preparing and selecting of HMPSA for transdermal use. HMPSA with different properties were prepared using styrene-isoprene-styrene triblock
Yong-Qi Fang et al.
Die Pharmazie, 67(2), 120-123 (2012-04-20)
beta-Asarone has significant pharmacological effects on the central nervous system. As a potential therapeutic agent to manage brain diseases, analysis of the pharmacokinetics of beta-asarone in brain is necessary. We used cardio-perfusion method to exclude the beta-asarone in the brain
Divyasree Sandeep et al.
Mutation research, 722(1), 62-68 (2011-03-29)
α-Asarone (1-propenyl-2,4,5-methoxybenzol), one of the active components of Acorus calamus extract, was examined for its efficacy as a radioprotector in mice exposed to lethal and sublethal whole-body γ-radiation. Oral administration of α-asarone 1h prior to the radiation exposure reduced radiation
Sreenath Nandakumar et al.
Biomedical chromatography : BMC, 27(3), 318-326 (2012-08-21)
β-Asarone (BAS), a phenylpropanoid from Acorus calamus Linn., has shown biological effects in the management of cognitive impairment conditions such as Alzheimer's disease. The present paper describes a selective and sensitive liquid chromatography-tandem mass spectrometric method (HPLC-MS/MS) using electrospray ionization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service