Skip to Content
Merck
All Photos(1)

Key Documents

11037

Sigma-Aldrich

Aluminum oxide hydrated

technical, ≥64% Al2O3 basis, powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Quality Level

grade

technical

form

powder

reaction suitability

core: aluminum
reagent type: catalyst

concentration

≥64% (Al2O3)

impurities

≤0.1% alkalis/earth alkalis (as SO4)

loss

≤35% loss on ignition, 1000 °C, 2 h

anion traces

sulfate (SO42-): ≤100 mg/kg

cation traces

Fe: ≤200 mg/kg

SMILES string

O[Al](O)O

InChI

1S/Al.3H2O/h;3*1H2/q+3;;;/p-3

InChI key

WNROFYMDJYEPJX-UHFFFAOYSA-K

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service