Skip to Content
Merck
All Photos(1)

Key Documents

09340

Sigma-Aldrich

2-Aminopyridine

purum, ≥98.0% (NT)

Synonym(s):

2-AP, 2-Pyridinamine, 2-Pyridylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2
CAS Number:
Molecular Weight:
94.11
Beilstein:
105785
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (NT)

bp

204-210 °C (lit.)

mp

54-58 °C (lit.)
54-59 °C

SMILES string

Nc1ccccn1

InChI

1S/C5H6N2/c6-5-3-1-2-4-7-5/h1-4H,(H2,6,7)

InChI key

ICSNLGPSRYBMBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to label neutral and acidic glycans for fluorescent and UV detection in HPLC. 2-AP has also been used to derivatize sialyloligosaccharides for detection in FAB-MS.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fotini N Lamari et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 793(1), 15-36 (2003-07-26)
Carbohydrates, either alone or as constituents of glycoproteins, proteoglycans and glycolipids, are mediators of several cellular events and (patho)physiological processes. Progress in the "glycome" project is closely related to the analytical tools used to define carbohydrate structure and correlate structure
M Okamoto
Bioscience, biotechnology, and biochemistry, 65(11), 2519-2527 (2002-01-17)
To establish a new protocol for sensitive detection and structural characterization of sialyl oligosaccharides, their sensitivities and structural information from mass spectrometry and tandem mass spectrometry with FAB-, ESI-, and MALDI were evaluated in detail. Among these ionization methods, FAB-MS
Philipp Ottiger et al.
The Journal of chemical physics, 136(17), 174308-174308 (2012-05-16)
The S(1)/S(2) state exciton splittings of symmetric doubly hydrogen-bonded gas-phase dimers provide spectroscopic benchmarks for the excited-state electronic couplings between UV chromophores. These have important implications for electronic energy transfer in multichromophoric systems ranging from photosynthetic light-harvesting antennae to photosynthetic
Daniela P Gouvêa et al.
Ultrasonics sonochemistry, 19(6), 1127-1131 (2012-04-10)
The efficient multicomponent synthesis of thiazolidinones from the reaction of arenealdehydes, mercaptoacetic acid and 2-picolilamine or 2-aminopyridine under ultrasound irradiation are reported. The reaction with 2-aminopyridine needs a Lewis acid catalysis to afford the corresponding 2-aryl-3-(pyridin-2-yl)-1,3-thiazolidin-4-ones. All novel compounds were
Honggen Wang et al.
Journal of the American Chemical Society, 132(38), 13217-13219 (2010-09-09)
A novel and efficient synthesis of pyrido[1,2-a]benzimidazoles through direct intramolecular aromatic C-H amination of N-aryl-2-aminopyridines has been developed. The reaction, cocatalyzed by Cu(OAc)(2) and Fe(NO(3))(3)·9H(2)O, is carried out in DMF under a dioxygen atmosphere. Diversified pyrido[1,2-a]benzimidazoles containing various substitution patterns

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Protocols

Explore mass spectrometry analysis of glycans for glycomic & glycoproteomic neutral & acidic glycan analysis. See a general mass spec glycan analysis procedure.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service