Skip to Content
Merck
All Photos(1)

Documents

09267

Sigma-Aldrich

(R)-3-Amino-1,2-propanediol

≥98.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H9NO2
CAS Number:
Molecular Weight:
91.11
Beilstein:
4290608
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (T)

optical activity

[α]20/D +28±1°, c = 4% in 5 M HCl

SMILES string

NC[C@@H](O)CO

InChI

1S/C3H9NO2/c4-1-3(6)2-5/h3,5-6H,1-2,4H2/t3-/m1/s1

InChI key

KQIGMPWTAHJUMN-GSVOUGTGSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Lider et al.
Proceedings of the National Academy of Sciences of the United States of America, 92(11), 5037-5041 (1995-05-23)
The activation of T cells by antigens or mitogens leads to the secretion of cytokines and enzymes that shape the inflammatory response. Among these molecular mediators of inflammation is a heparanase enzyme that degrades the heparan sulfate scaffold of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service