Skip to Content
Merck
All Photos(1)

Key Documents

09180

Sigma-Aldrich

3-Amino-4-hydroxybenzenesulfonic acid

technical, ≥95% (T)

Synonym(s):

2-Aminophenol-4-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(OH)SO3H
CAS Number:
Molecular Weight:
189.19
Beilstein:
514765
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥95% (T)

ign. residue

≤2%

mp

≥300 °C (lit.)

functional group

sulfonic acid

SMILES string

Nc1cc(ccc1O)S(O)(=O)=O

InChI

1S/C6H7NO4S/c7-5-3-4(12(9,10)11)1-2-6(5)8/h1-3,8H,7H2,(H,9,10,11)

InChI key

ULUIMLJNTCECJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jolanta Polak et al.
New biotechnology, 33(2), 255-262 (2015-10-24)
This study demonstrates the optimisation of the main parameters of the laccase-mediated biosynthesis of high-intensity-coloured orange phenoxazine compound, 2-amino-3-oxo-3H-phenoxazine-8-sulfonic acid, and the antioxidative and dyeing properties. Among optimised parameters were the pH value, the activity of laccase, and the high
Milena Cojic et al.
Frontiers in endocrinology, 12, 610893-610893 (2021-09-08)
Vitamin D deficiency could play an important role in the pathogenesis of type 2 diabetes mellitus (T2DM) as it may alter several crucial processes in the development of diabetes and its complications, such as pancreatic insulin secretion, peripheral insulin resistance
Maureen Keller-Wood et al.
Pediatric research, 69(6), 491-496 (2011-03-08)
In the human and ovine fetus, the presence of 11β-hydroxysteroid dehydrogenase 1 allows cortisol and other corticosteroids to act at mineralocorticoid receptors (MRs) in lung and brain. To test the physiologic role of MRs in the late gestation fetus, fetal
Matthew A Bailey et al.
Hypertension (Dallas, Tex. : 1979), 54(4), 890-896 (2009-07-29)
Experiments in Cushing patients and healthy control subjects receiving adrenocorticotropic hormone (ACTH) indicate that transient renal sodium retention may contribute to the generation of hypertension. Here we have investigated the effect of chronic ACTH infusion on renal sodium handling in
Frances E Jones et al.
Disease models & mechanisms, 9(2), 165-176 (2016-02-04)
Collagen IV is a major component of basement membranes, and mutations in COL4A1, which encodes collagen IV alpha chain 1, cause a multisystemic disease encompassing cerebrovascular, eye and kidney defects. However, COL4A1 renal disease remains poorly characterized and its pathomolecular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service