Skip to Content
Merck
All Photos(2)

Key Documents

0908-BP

Sigma-Aldrich

Neu5Acα6`(6-su)LN-C3-PAA-biot

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
€968.00

€968.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
0.5 MG
€968.00

About This Item

UNSPSC Code:
12352201
NACRES:
NA.25

€968.00


Estimated to ship on22 May 2025


Request a Bulk Order

carrier

PAA carrier

conjugate

biotin conjugate

product line

BP-Series

Assay

95% (H-NMR)

form

lyophilized powder

mol wt

~20,000 Da

manufacturer/tradename

GlycoNZ

extent of labeling

20 mol% Biotin ligand density

technique(s)

flow cytometry: suitable
glycol-bead construction: suitable
glycol-surface construction: suitable

probe type

trisaccharide

probe presentation

polymeric

application(s)

histochemistry

storage temp.

−20°C

Related Categories

General description

BP-Series: Biotinylated Polymeric Probes
This series of glycoconjugate probes includes HOCH2CH2NH3+-salts for acidic sugars. The affinity of these probes is 102 to 105 times higher than that of the corresponding free sugars. The spacer-arm is normally –(CH2)3–, while the spacer arm for biotin is –(CH2)6–. The flexible polymer chain behaves as an additional spacer.

Used as a high molecular weight carrier, poly[N-(2-hydroxyethyl)acrylamide], abbreviated as PAA, has
low non-specific sorption, and is stable to chemical and proteolytic action.

Application

Glycoconjugates are used to study the functions of carbohydrates, carbohydrate-binding proteins in biological processes, and on the discovery of new carbohydrate-mediated interactions.

Preparation Note

Purity: synthetic ω-aminoalkyl glycosides used for coupling with the polymer have a purity of >95% (HPLC and 1H-NMR)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service