Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
H2NC6H3(NO2)OH
CAS Number:
Molecular Weight:
154.12
EC Number:
202-767-9
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
776533
MDL number:
assay
≥99.0% (NT)
mp
140-143 °C (lit.), 143-145 °C
SMILES string
Nc1cc(ccc1O)[N+]([O-])=O
InChI
1S/C6H6N2O3/c7-5-3-4(8(10)11)1-2-6(5)9/h1-3,9H,7H2
InChI key
VLZVIIYRNMWPSN-UHFFFAOYSA-N
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
212.0 °F - closed cup
flash_point_c
100 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
