Skip to Content
Merck

Skip To

08375

Ethyl hexanoate

analytical standard

Synonym(s):

Caproic acid ethyl ester, Ethyl caproate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mL
Check Cart for Availability
€ 69,70

About This Item

Linear Formula:
CH3(CH2)4COOC2H5
CAS Number:
Molecular Weight:
144.21
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
EC Number:
204-640-3
Beilstein/REAXYS Number:
1701293
MDL number:

€ 69,70


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor density

5 (vs air)

assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.2% water

refractive index

n20/D 1.406-1.408, n20/D 1.407 (lit.)

bp

168 °C (lit.)

density

0.869 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCC(=O)OCC

InChI

1S/C8H16O2/c1-3-5-6-7-8(9)10-4-2/h3-7H2,1-2H3

InChI key

SHZIWNPUGXLXDT-UHFFFAOYSA-N

General description

Ethyl hexanoate, an ester, is a volatile aroma and fragrance compound which impacts the quality and flavor of wine, beer and Japanese sake.[1]

Application

Ethyl hexanoate may be used as an analytical reference standard for the determination of the analyte in wine and beer samples by chromatography based techniques.[1][2]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W243906W243914W270814
grade

analytical standard

grade

FG, Halal, Kosher

grade

FG, Halal, Kosher, natural

grade

FG, Halal, Kosher, natural

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

flavors and fragrances

application(s)

flavors and fragrances

application(s)

flavors and fragrances

assay

≥99.0% (GC)

assay

≥98%

assay

≥98%

assay

≥99%

format

neat

format

-

format

-

format

-

density

0.869 g/mL at 25 °C (lit.)

density

0.869 g/mL at 25 °C (lit.)

density

0.869 g/mL at 25 °C (lit.)

density

0.885 g/mL at 25 °C (lit.)

impurities

≤0.2% water

impurities

-

impurities

-

impurities

-


Still not finding the right product?

Explore all of our products under Ethyl hexanoate


pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

127.4 °F - closed cup

flash_point_c

53 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters