Skip to Content
Merck
All Photos(2)

Key Documents

05846

Supelco

Iscotrizinol

analytical standard

Synonym(s):

4,4-[[6-[[4-[[(1,1-Dimethylethyl)amino]carbonyl]phenyl]amino]-1,3,5-triazine-2,4-diyl]diimino]bisbenzoic acid 1,1′-bis(2-ethylhexyl) ester, 4,6-Bis[4-(2-ethylhexyloxycarbonyl)anilino]-2-[4-(tert-butylaminocarbonyl)anilino]-1,3,5-triazine, Bis(2-ethylhexyl) 4,4‘-[(6-{[4-(tert-butylcarbamoyl)phenyl]amino}-1,3,5-triazine-2,4-diyl)diimino]dibenzoate, Diethylhexyl butamido triazone

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€370.00

€370.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€370.00

About This Item

Empirical Formula (Hill Notation):
C44H59N7O5
CAS Number:
Molecular Weight:
765.98
Beilstein:
11322517
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€370.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCC(CC)COC(=O)c1ccc(Nc2nc(Nc3ccc(cc3)C(=O)NC(C)(C)C)nc(Nc4ccc(cc4)C(=O)OCC(CC)CCCC)n2)cc1

InChI

1S/C44H59N7O5/c1-8-12-14-30(10-3)28-55-39(53)33-18-24-36(25-19-33)46-42-48-41(45-35-22-16-32(17-23-35)38(52)51-44(5,6)7)49-43(50-42)47-37-26-20-34(21-27-37)40(54)56-29-31(11-4)15-13-9-2/h16-27,30-31H,8-15,28-29H2,1-7H3,(H,51,52)(H3,45,46,47,48,49,50)

InChI key

OSCJHTSDLYVCQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iscotrizinol is an organic chemical ultraviolet (UV) filter approved for use in sunscreens to absorb UVB and some UVA radiations.[1][2]

Application

Iscotrizinol may be used as a reference material for the determination of the analyte in the marine sediments by liquid chromatography/tandem mass spectrometry (LC-MS/MS).[3]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sunscreen use: controversies, challenges and regulatory aspects
Loden M, et al.
British Journal of Dermatology, 165(2), 255-262 (2011)
Environmental occurrence and distribution of organic UV stabilizers and UV filters in the sediment of Chinese Bohai and Yellow Seas
Apel C, et al.
Environmental Pollution (Barking, Essex : 1987), 235, 85-94 (2018)
Trade-offs in ecosystem impacts from nanomaterial versus organic chemical ultraviolet filters in sunscreens
Hanigan D, et al.
Water Research, 139, 281-290 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service