Skip to Content
Merck
All Photos(1)

Key Documents

05828

Supelco

Isopropyl 4-hydroxybenzoate

analytical standard

Synonym(s):

4-Hydroxybenzoic acid isopropyl ester, Isopropyl paraben

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€65.40

€65.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€65.40

About This Item

Empirical Formula (Hill Notation):
C10H12O3
CAS Number:
Molecular Weight:
180.20
Beilstein:
2641564
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€65.40


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

O=C(OC(C)C)C1=CC=C(O)C=C1

InChI

1S/C10H12O3/c1-7(2)13-10(12)8-3-5-9(11)6-4-8/h3-7,11H,1-2H3

InChI key

CMHMMKSPYOOVGI-UHFFFAOYSA-N

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Pastor-Belda et al.
Journal of chromatography. A, 1564, 102-109 (2018-06-17)
This study describes the use of a magnetic multi-walled carbon nanotube composite for the preconcentration of nine parabens (methyl, ethyl, propyl, isopropyl, butyl, isobutyl, pentyl, phenyl and benzylparaben) from water and urine samples after in situ acetylation. The enriched extracts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service