04907
N,N-Bis-[(R)-1-phenylethyl]dibenzo[d,f][1,3,2]dioxaphosphepin-6-amine
≥99.0%
Synonym(s):
(R,R)-N-(5,7-Diox-6-phosphadibenzo[a,c]cyclohepten-6-yl)bis(1-phenylethyl)amine, O,O′-(2,2′-Biphenyldiyl) N,N-bis[(R)-1-phenylethyl]phosphoramidite, BIPOL-A1(R)
About This Item
Quality Level
Assay
≥99.0%
optical purity
enantiomeric ratio: ≥97.0:3.0
reaction suitability
reaction type: click chemistry
mp
99-102 °C
functional group
amine
phenyl
SMILES string
C[C@@H](N([C@H](C)c1ccccc1)P2Oc3ccccc3-c4ccccc4O2)c5ccccc5
InChI
1S/C28H26NO2P/c1-21(23-13-5-3-6-14-23)29(22(2)24-15-7-4-8-16-24)32-30-27-19-11-9-17-25(27)26-18-10-12-20-28(26)31-32/h3-22H,1-2H3
InChI key
JISGHECLGYELKD-UHFFFAOYSA-N
Application
- In the asymmetric hydrovinylation of 1,3-diene.
- In the 1,4-asymmetric conjugate addition of 3-substituted cyclohexenones catalyzed by copper.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service