Skip to Content
Merck
All Photos(1)

Key Documents

04650590

Hesperidin

primary reference standard

Synonym(s):

3′,5,7-Trihydroxy 4′-methoxyflavanone 7-rutinoside, Hesperetin 7-rhamnoglucoside, Hesperitin-7-rutinoside

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€309.00

€309.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€309.00

About This Item

Empirical Formula (Hill Notation):
C28H34O15
CAS Number:
Molecular Weight:
610.56
Beilstein:
75140
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€309.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

mp

250-255 °C (dec.) (lit.)

application(s)

food and beverages

SMILES string

COc1ccc(cc1O)[C@@H]2CC(=O)c3c(O)cc(O[C@@H]4O[C@H](CO[C@@H]5O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)[C@@H](O)[C@H](O)[C@H]4O)cc3O2

InChI

1S/C28H34O15/c1-10-21(32)23(34)25(36)27(40-10)39-9-19-22(33)24(35)26(37)28(43-19)41-12-6-14(30)20-15(31)8-17(42-18(20)7-12)11-3-4-16(38-2)13(29)5-11/h3-7,10,17,19,21-30,32-37H,8-9H2,1-2H3/t10-,17-,19+,21-,22+,23+,24-,25+,26+,27+,28+/m0/s1

InChI key

QUQPHWDTPGMPEX-QJBIFVCTSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products
Studies of the behavior of hesperidin and hesperetin (the aglycone) with DMPC liposomes find the aglycone penetrates more deeply and binds more strongly to the "membrane." This may explain the better bioavailability of bioflavonoid aglycones in general.

Biochem/physiol Actions

Flavonoid phytochemical extracted from citrus species. It is the rhamnoglucoside (rutinoside) of hesperetin.

Other Notes

This compound is commonly found in plants of the genus: angelica mentha valeriana

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Gowrikumar Saiprasad et al.
European journal of cancer (Oxford, England : 1990), 50(14), 2489-2507 (2014-07-23)
Abnormalities in the homeostasis mechanisms involved in cell survival and apoptosis are contributing factors for colon carcinogenesis. Interventions of these mechanisms by pharmacologically safer agents gain predominance in colon cancer prevention. We previously reported the chemopreventive efficacy of hesperidin against
Asjad Visnagri et al.
Pharmaceutical biology, 52(7), 814-828 (2014-02-25)
Diabetic neuropathy (DN) is one of the most common long-term complications of diabetes mellitus and clinically can be characterized by an elevated nociceptive response with electrophysiological conduction abnormalities. The present investigation was designed to evaluate the neuroprotective effect of hesperidin
Silvia Yumnam et al.
PloS one, 9(6), e101321-e101321 (2014-07-01)
Hesperidin, a natural flavonoid abundantly present in Citrus is known for its anti-cancer, anti-oxidant and anti-inflammatory properties. In this study we examined the effect of hesperidin on HepG2 cells. HepG2 cells treated with various concentration of hesperidin undergo a distinct
G Allam et al.
Journal of helminthology, 88(3), 362-370 (2013-05-10)
Hesperidin has been reported to exert a wide range of pharmacological effects, including antifungal, antiviral, antioxidant, anti-inflammatory and anticarcinogenic activities. Herein, the schistosomicidal activity of this compound was evaluated in vitro and in vivo. Using an in vitro assay, a
Feng-Quan Tan et al.
BMC plant biology, 15, 89-89 (2015-04-08)
Polyploidy has often been considered to confer plants a better adaptation to environmental stresses. Tetraploid citrus rootstocks are expected to have stronger stress tolerance than diploid. Plenty of doubled diploid citrus plants were exploited from diploid species for citrus rootstock

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service