Skip to Content
Merck
All Photos(1)

Key Documents

04383

Sigma-Aldrich

1-Butyl-2,3-dimethylimidazolium tetrafluoroborate

for electrochemistry, ≥99.0% (HPLC/T)

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€244.00

€244.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
€244.00

About This Item

Empirical Formula (Hill Notation):
C9H17BF4N2
CAS Number:
Molecular Weight:
240.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€244.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥99.0% (HPLC/T)

impurities

≤0.3% water
≤50 mg/kg halogens (as chloride)

mp

37 °C

density

1.198 g/mL at 20 °C (lit.)

SMILES string

F[B-](F)(F)F.CCCCn1cc[n+](C)c1C

InChI

1S/C9H17N2.BF4/c1-4-5-6-11-8-7-10(3)9(11)2;2-1(3,4)5/h7-8H,4-6H2,1-3H3;/q+1;-1

InChI key

VCAIYEJBOWHUGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Butyl-2,3-dimethylimidazolium tetrafluoroborate can be used as a component of graphene nanoribbon and cobalt phthalocyanine based nanocomposites modified electrode, which is applicable in the determination of pesticide fenobucarb in vegetables.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A highly sensitive fenobucarb electrochemical sensor based on graphene nanoribbons-ionic liquid-cobalt phthalocyanine composites modified on screen-printed carbon electrode coupled with a flow injection analysis
Kunpatee K, et al.
Journal of Electroanalytical Chemistry, 855, 113630-113630 (2019)

Articles

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service