Skip to Content
Merck
All Photos(1)

Key Documents

04340

Supelco

3-(N-Ethyl-3-methylanilino)-2-hydroxypropanesulfonic acid sodium salt

for enzymic, spectrophotometric determination of H2O2, ≥98.0%

Synonym(s):

3-(N-Ethyl-m-toluidino)-2-hydroxypropanesulfonic acid sodium salt, N-Ethyl-N-(2-hydroxy-3-sulfopropyl)-m-toluidine sodium salt, Sodium 3-(N-ethyl-3-methylanilino)-2-hydroxypropanesulfonate, Sodium N-ethyl-N-(2-hydroxy-3-sulfopropyl)-m-toluidine, TOOS

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€98.00

€98.00


Estimated to ship on13 April 2025


Request a Bulk Order

Select a Size

Change View
500 MG
€98.00

About This Item

Empirical Formula (Hill Notation):
C12H18NNaO4S
CAS Number:
Molecular Weight:
295.33
Beilstein:
5689521
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

€98.00


Estimated to ship on13 April 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (dried material, NT)
≥98.0%

form

powder or crystals

quality

for enzymic, spectrophotometric determination of H2O2

technique(s)

UV/Vis spectroscopy: suitable

impurities

8-12% water

SMILES string

[Na+].CCN(CC(O)CS([O-])(=O)=O)c1cccc(C)c1

InChI

1S/C12H19NO4S.Na/c1-3-13(8-12(14)9-18(15,16)17)11-6-4-5-10(2)7-11;/h4-7,12,14H,3,8-9H2,1-2H3,(H,15,16,17);/q;+1/p-1

InChI key

IRQRBVOQGUPTLG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Water-soluble reagent for the enzymatic photometric determination of hydrogen peroxide[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamaoku, K., et al.
Chemical & Pharmaceutical Bulletin, 30, 2492-2492 (1982)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service