Skip to Content
Merck
All Photos(1)

Key Documents

04061

Sigma-Aldrich

1-(Fmoc-amino)cyclohexanecarboxylic acid

≥98.0% (T)

Synonym(s):

Fmoc-homocycloleucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23NO4
CAS Number:
Molecular Weight:
365.42
Beilstein:
7887646
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98.0% (T)

form

powder

color

white

mp

185-187 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)C1(CCCCC1)NC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C22H23NO4/c24-20(25)22(12-6-1-7-13-22)23-21(26)27-14-19-17-10-4-2-8-15(17)16-9-3-5-11-18(16)19/h2-5,8-11,19H,1,6-7,12-14H2,(H,23,26)(H,24,25)

InChI key

VCIVAWBKUQJNSX-UHFFFAOYSA-N

Other Notes

Unnatural amino acids employed for peptide libraries.[1][2]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Lescrinier et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 4, 425-425 (1998)
A.P. Combs et al.
Journal of the American Chemical Society, 118, 287-287 (1996)
Anil Zechariah et al.
Stroke, 44(6), 1690-1697 (2013-05-02)
Therapeutic angiogenesis aims at improving cerebral blood flow by amplification of vascular sprouting, thus promoting tissue survival under conditions of subsequent ischemia. It remains unknown whether induced angiogenesis leads to the formation of functional vessels that indeed result in hemodynamic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service