Skip to Content
Merck
All Photos(1)

Key Documents

03761

Sigma-Aldrich

Fumaric acid

tested according to USP/NF

Synonym(s):

(2E)-2-Butenedioic acid, trans-Butenedioic acid, Acidum fumaricum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH=CHCOOH
CAS Number:
Molecular Weight:
116.07
Beilstein:
605763
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021709
PubChem Substance ID:
NACRES:
NA.21

Agency

tested according to USP/NF

Quality Level

vapor pressure

1.7 mmHg ( 165 °C)

Assay

≥99.5%

form

fine crystals
powder

autoignition temp.

1364 °F

expl. lim.

40 %

mp

298-300 °C (subl.) (lit.)

application(s)

pharmaceutical (small molecule)

functional group

carboxylic acid

SMILES string

OC(=O)\C=C\C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+

InChI key

VZCYOOQTPOCHFL-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fumaric acid or trans-butenedioic acid, is crystalline compound that occurs widely in nature.

Application

Fumaric acid may be used in the preparation of L-Lysine-fumaric acid crystals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

523.4 °F

Flash Point(C)

273 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

UV-curable methacrylated/fumaric acid modified epoxy as a potential support for enzyme immobilization.
Kahraman MV, et al.
Reactive functional Polymers, 67(2), 97-103 (2007)
C J Newbold et al.
The British journal of nutrition, 94(1), 27-35 (2005-08-24)
Fifteen potential precursors of propionate were tested for their ability to decrease CH4 production by ruminal fluid in vitro. Sodium acrylate and sodium fumarate produced the most consistent effects in batch cultures, with 50 % of the added precursors being
Comparison of dilute mineral and organic acid pretreatment for enzymatic hydrolysis of wheat straw.
Kootstra AMJ, et al.
Biochemical Engineering Journal, 46(2), 126-131 (2009)
Vibrational spectra and second harmonic generation in molecular complexes of L-lysine with L-tartaric, D, L-malic, acetic, arsenous, and fumaric acids.
Marchewka MK, et al.
Crystal Growth & Design, 3(4), 587-592 (2003)
N Cao et al.
Applied and environmental microbiology, 62(8), 2926-2931 (1996-08-01)
An integrated system of simultaneous fermentation-adsorption for the production and recovery of fumaric acid from glucose by Rhizopus oryzae was investigated. The system was constructed such that growing Rhizopus mycelia were self-immobilized on the plastic discs of a rotary biofilm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service