Skip to Content
Merck
All Photos(1)

Documents

03661

Sigma-Aldrich

Boc-β-Homomet-OH

≥98.0% (TLC)

Synonym(s):

(R)-3-(Boc-amino)-5-(methylthio)pentanoic acid, Boc-L-β-homomethionine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21NO4S
CAS Number:
Molecular Weight:
263.35
Beilstein:
8319678
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (TLC)

form

powder with small lumps

color

white

mp

81-83 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CSCC[C@@H](CC(O)=O)NC(=O)OC(C)(C)C

InChI

1S/C11H21NO4S/c1-11(2,3)16-10(15)12-8(5-6-17-4)7-9(13)14/h8H,5-7H2,1-4H3,(H,12,15)(H,13,14)/t8-/m0/s1

InChI key

HBFKPNZCUUZRLA-QMMMGPOBSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service