Skip to Content
Merck
All Photos(1)

Key Documents

03320585

Carminic acid

primary reference standard

Synonym(s):

Natural Red 4

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€256.00

€256.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€256.00

About This Item

Empirical Formula (Hill Notation):
C22H20O13
CAS Number:
Molecular Weight:
492.39
Colour Index Number:
75470
Beilstein:
71651
EC Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.24

€256.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

SMILES string

Cc1c(C(O)=O)c(O)cc2C(=O)c3c(O)c(O)c([C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)c(O)c3C(=O)c12

InChI

1S/C22H20O13/c1-4-8-5(2-6(24)9(4)22(33)34)13(25)10-11(15(8)27)16(28)12(18(30)17(10)29)21-20(32)19(31)14(26)7(3-23)35-21/h2,7,14,19-21,23-24,26,28-32H,3H2,1H3,(H,33,34)/t7-,14-,19+,20-,21+/m1/s1

InChI key

DGQLVPJVXFOQEV-JNVSTXMASA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service