Skip to Content
Merck
All Photos(1)

Documents

03128

Supelco

Diethylene glycol

analytical standard

Synonym(s):

2,2′-Oxydiethanol, 2-Hydroxyethyl ether, Bis(2-hydroxyethyl) ether, Diglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)2O
CAS Number:
Molecular Weight:
106.12
Beilstein:
969209
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.14 (vs air)

vapor pressure

0.01 mmHg ( 20 °C)

description

non-ionic

Assay

≥99.5% (GC)

form

viscous liquid (clear)

autoignition temp.

442 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

2-12.3 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.447 (lit.)
n20/D 1.447

bp

245 °C (lit.)

mp

−10 °C (lit.)

density

1.115-1.117 g/mL at 20 °C
1.118 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

OCCOCCO

InChI

1S/C4H10O3/c5-1-3-7-4-2-6/h5-6H,1-4H2

InChI key

MTHSVFCYNBDYFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

Diethylene glycol may be used as an analytical reference standard for the determination of the analyte in toothpaste, poly(ethylene terephthalate) yarns, pharmaceutical products and human biological samples by various chromatography techniques.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation of diethylene glycol as a side reaction during production of polyethylene terephthalate.
Hovenkamp, S. G., and J. P. Munting.
Journal of Polymer Science. Part A-1, Polymer Chemistry, 8.3, 679-682 (1970)
Fast determination of toxic diethylene glycol in toothpaste by ultra-performance liquid chromatography-time of flight mass spectrometry
Hernandez F, et al.
Analytical and Bioanalytical Chemistry, 391(3), 1021-1021 (2008)
Validated gas chromatographic-mass spectrometric assay for determination of the antifreezes ethylene glycol and diethylene glycol in human plasma after microwave-assisted pivalylation
Maurer HH, et al.
Journal of Chromatography. B, Biomedical Applications, 754(2), 401-409 (2001)
Identification and quantification of ethylene glycol and diethylene glycol in plasma using gas chromatography-mass spectrometry
Maurer Hans and Kessler C
Archives of Toxicology, 62(1), 66-69 (1988)
Gas chromatographic determination of diethylene glycol in poly (ethylene terephthalate)
Ponder LH
Analytical Chemistry, 40(1), 229-231 (1968)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service