Skip to Content
Merck
All Photos(1)

Key Documents

02977

Sigma-Aldrich

2-(4-Isothiocyanatophenoxy)ethyl tosylate

≥98.0% (TLC)

Synonym(s):

2-(4-Isothiocyanatophenoxy)ethyl p-toluenesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15NO4S2
CAS Number:
Molecular Weight:
349.42
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (TLC)

mp

108-112 °C

SMILES string

Cc1ccc(cc1)S(=O)(=O)OCCOc2ccc(cc2)N=C=S

InChI

1S/C16H15NO4S2/c1-13-2-8-16(9-3-13)23(18,19)21-11-10-20-15-6-4-14(5-7-15)17-12-22/h2-9H,10-11H2,1H3

InChI key

LILJDLJORCAIDG-UHFFFAOYSA-N

Application

Hetero-bifunctional linker specially for the quaternisation of pyridyl-nitrogen in order to introduce an amine or thiol reactive group. Synthetic building block for the preparation of fluorescent labels[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Litak, P.T.;Kauffman, J.M.
Journal of Heterocyclic Chemistry, 31, 457-457 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service