Skip to Content
Merck
All Photos(2)

Documents

01354

Supelco

Quinoline Yellow 2SF

analytical standard

Synonym(s):

2-(2-Quinolinyl)-1H-indene-1,3(2H)-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H11NO2
CAS Number:
Molecular Weight:
273.29
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

O=C1C2=C(C=CC=C2)C(C1C3=NC4=CC=CC=C4C=C3)=O

InChI

1S/C18H11NO2/c20-17-12-6-2-3-7-13(12)18(21)16(17)15-10-9-11-5-1-4-8-14(11)19-15/h1-10,16H

InChI key

IZMJMCDDWKSTTK-UHFFFAOYSA-N

General description

Quinoline Yellow 2SF is a greenish-yellow dye, widely used as a coloring agent for paints, wax, wools, etc. It is easy to synthesize and highly soluble.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tandem malonate-based glucosides (TMGs) for membrane protein structural studies
Hussain H, et al.
Scientific reports, 7(1), 1-11 (2017)
Ruifang Qi et al.
The Analyst, 139(23), 6168-6177 (2014-10-15)
A synthetic protocol for the preparation of an indium oxide nanoparticle-functionalized poly(methacrylic acid-glycidyl methacrylate-ethylene dimethacrylate-ethanediamine) monolithic column is reported. Various techniques, including scanning electron microscopy, transmission electron microscopy, Fourier transform infrared spectroscopy, X-ray photoelectron spectroscopy, and thermal gravimetric analysis-derivative thermogravimetric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service