Skip to Content
Merck
All Photos(1)

Key Documents

01050595

Eugenol

primary reference standard

Synonym(s):

2-Methoxy-4-(2-propenyl)phenol, 4-Allyl-2-methoxyphenol, 4-Allylguaiacol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(H2C=CHCH2)C6H3-2-(OCH3)OH
CAS Number:
Molecular Weight:
164.20
Beilstein:
1366759
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

form

liquid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

refractive index

n20/D 1.541 (lit.)

bp

254 °C (lit.)

mp

−12-−10 °C (lit.)

density

1.067 g/mL at 25 °C (lit.)

application(s)

food and beverages

shipped in

dry ice

storage temp.

−20°C

SMILES string

COc1cc(CC=C)ccc1O

InChI

1S/C10H12O2/c1-3-4-8-5-6-9(11)10(7-8)12-2/h3,5-7,11H,1,4H2,2H3

InChI key

RRAFCDWBNXTKKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

255.2 °F - closed cup

Flash Point(C)

124 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

K Markowitz et al.
Oral surgery, oral medicine, and oral pathology, 73(6), 729-737 (1992-06-01)
Eugenol-containing dental materials are frequently used in clinical dentistry. When zinc oxide-eugenol (ZOE) is applied to a dentinal cavity, small quantities of eugenol diffuse through the dentin to the pulp. Low concentrations of eugenol exert anti-inflammatory and local anesthetic effects
W R Hume
Journal of oral rehabilitation, 21(4), 469-473 (1994-07-01)
The diffusion gradient which develops across dentine and the clearance into the capillary circulation at the pulpal side of the tissue both reduce the concentration of potential toxins applied to dentine, thus protecting pulpal cells. The data presented on eugenol
Guy P Kamatou et al.
Molecules (Basel, Switzerland), 17(6), 6953-6981 (2012-06-26)
Eugenol is a major volatile constituent of clove essential oil obtained through hydrodistillation of mainly Eugenia caryophyllata (=Syzygium aromaticum) buds and leaves. It is a remarkably versatile molecule incorporated as a functional ingredient in numerous products and has found application
Ali Fulcher et al.
Journal of medical entomology, 51(4), 896-899 (2014-08-15)
The effect of spraying a mixture of the insect growth regulator (IGR) pyriproxyfen (1 mg/liter) and either 1% boric acid sugar bait or eugenol sugar bait on croton petra plants (Codiaeum variegatum L.) was evaluated against the container-inhabiting mosquito, Aedes
Roger I Vargas et al.
Vitamins and hormones, 83, 575-595 (2010-09-14)
Worldwide, an important aspect of invasive insect pest management is more effective, safer detection and control systems. Phenyl propanoids are attractive to numerous species of Dacinae fruit flies. Methyl eugenol (ME) (4-allyl-1, 2-dimethoxybenzene-carboxylate), cue-lure (C-L) (4-(p-acetoxyphenyl)-2-butanone), and raspberry ketone (RK)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service