Skip to Content
Merck
All Photos(1)

Key Documents

00899

Supelco

Khellin

analytical standard

Synonym(s):

4,9-Dimethoxy-7-methyl-5H-furo[3,2-g][1]benzopyran-5-one

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€175.00

€175.00


Estimated to ship on13 April 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€175.00

About This Item

Empirical Formula (Hill Notation):
C14H12O5
CAS Number:
Molecular Weight:
260.24
Beilstein:
263185
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€175.00


Estimated to ship on13 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

bp

180-200 °C/0.05 mmHg (lit.)

mp

150-154 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

COc1c2OC(C)=CC(=O)c2c(OC)c3ccoc13

InChI

1S/C14H12O5/c1-7-6-9(15)10-11(16-2)8-4-5-18-12(8)14(17-3)13(10)19-7/h4-6H,1-3H3

InChI key

HSMPDPBYAYSOBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Khellin, is reported to be an efficient drug for treating vitiligo when combined with ultraviolet A irradiation.[1]

Application

Khellin has been used as a reference standard for supercritical fluid extraction of khellin from Ammi visnaga L. fruits to understand the various medicinal effects of γ-Pyrones by reversed phase HPLC method.[2]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phototherapeutic, photobiologic, and photosensitizing properties of khellin.
Morliere P, et al.
The Journal of Investigative Dermatology, 90(5), 720-724 (1988)
Supercritical fluid extraction of γ-Pyrones from Ammi visnaga L. fruits.
Bishr M, et al
Future Journal of Pharmaceutical Sciences (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service