Skip to Content
Merck
All Photos(1)

Documents

00796

Sigma-Aldrich

1-Ethyl-3-methylimidazolium dicyanamide

≥98.5% (HPLC/T)

Synonym(s):

EMIM DCA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11N5
CAS Number:
Molecular Weight:
177.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.5% (HPLC/T)

impurities

≤0.5% water

SMILES string

N#C[N-]C#N.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.C2N3/c1-3-8-5-4-7(2)6-8;3-1-5-2-4/h4-6H,3H2,1-2H3;/q+1;-1

InChI key

MKHFCTXNDRMIDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Ethyl-3-methylimidazolium dicyanamide (EMIM-DCA) is an ionic liquid that is often used as a solvent in various chemical reactions and as an electrolyte in batteries and capacitors. It is considered a greener alternative to organic traditional solvents due to its non-toxicity.

Application

1-Ethyl-3-methylimidazolium dicyanamide can be used in the development of polyvinyl alcohol based polymeric membranes for the for the effective separation of CO2/H2.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

429.8 °F - closed cup

Flash Point(C)

221 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service