Skip to Content
Merck
All Photos(1)

Key Documents

00790

Sigma-Aldrich

Acetophenone

puriss. p.a., ≥99.0% (GC)

Synonym(s):

Methyl phenyl ketone

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€64.10

€64.10


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€64.10

About This Item

Linear Formula:
CH3COC6H5
CAS Number:
Molecular Weight:
120.15
Beilstein:
605842
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39023804
PubChem Substance ID:
NACRES:
NA.21

€64.10


Estimated to ship on30 May 2025


Request a Bulk Order

vapor density

4.1 (vs air)

Quality Level

vapor pressure

0.45 mmHg ( 25 °C)
1 mmHg ( 15 °C)

grade

puriss. p.a.

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

1058 °F

impurities

≤0.02% free acid (as CH3COOH)
≤0.3% water

refractive index

n20/D 1.534 (lit.)
n20/D 1.535

bp

202 °C (lit.)

mp

19-20 °C (lit.)

transition temp

solidification point 19-20 °C

density

1.03 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

functional group

ketone
phenyl

SMILES string

CC(=O)c1ccccc1

InChI

1S/C8H8O/c1-7(9)8-5-3-2-4-6-8/h2-6H,1H3

InChI key

KWOLFJPFCHCOCG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetophenone (Methyl phenyl ketone) is an alkyl phenyl ketone. Its triplet absorption spectra in non-polar solvents has been evaluated by nanosecond laser photolysis.[1] Photoreduction of acetophenone has been investigated.[2] N-chelate ligands based on chiral oxazolines catalyzed enantioselective hydrosilylation of acetophenone with α-naphtylphenylsilane has been reported.[3] Intersystem crossing of aromatic aldehydes and ketones (benzaldehyde, acetophenone and benzophenone) has been studied.[4] Rhodium(I) complexes containing optically active 2-(2′-pyridyl)pyridines efficiently catalyzes the reduction of acetophenone.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optically active nitrogen ligans: III. Enantioface-discriminating transfer hydrogenation of acetophenone catalyzed by rhodium (I) complexes with chiral 2-(2'-pyridyl) pyridines.
Botteghi C, et al.
Journal of Organometallic Chemistry, 304(1), 217-225 (1986)
Comment on "closing the loop on bond selective chemistry using tailored strong field laser pulses".
Xin Zhu et al.
The journal of physical chemistry. A, 113(17), 5264-5266 (2009-04-24)
Effects of solvent and substituents on the absorption spectra of triplet acetophenone and the acetophenone ketyl radical studied by nanosecond laser photolysis.
Lutz H, et al.
The Journal of Physical Chemistry, 77(14), 1758-1762 (1973)
Photoreduction of acetophenone and substituted acetophenones.
Yang N-C, et al.
Journal of the American Chemical Society, 89(21), 5466-5468 (1967)
Enantioselective hydrosilylation of acetophenone with rhodium/oxazolines catalysts.
Balavoine G, et al.
Tetrahedron Letters, 30(38), 5141-5144 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service