Skip to Content
Merck
All Photos(1)

Key Documents

00469

Sigma-Aldrich

α-Ionol

technical, ≥90% (GC)

Synonym(s):

4-(2,6,6-Trimethyl-2-cyclohexenyl)-3-buten-2-ol

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€50.80

€50.80


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€50.80

About This Item

Empirical Formula (Hill Notation):
C13H22O
CAS Number:
Molecular Weight:
194.31
Beilstein:
2327829
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€50.80


Estimated to ship on21 May 2025


Request a Bulk Order

grade

technical

Quality Level

Assay

≥90% (GC)

form

liquid

refractive index

n20/D 1.491

bp

127 °C/15 mmHg (lit.)

density

0.918 g/mL at 20 °C (lit.)

functional group

hydroxyl

SMILES string

CC(O)\C=C\C1C(C)=CCCC1(C)C

InChI

1S/C13H22O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h6-8,11-12,14H,5,9H2,1-4H3/b8-7+

InChI key

PWDOJWCZWKWKSE-BQYQJAHWSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grant T McQuate et al.
Insects, 9(2) (2018-05-05)
Bactrocera latifrons (Hendel) (Diptera: Tephritidae) is a pest fruit fly species native to Oriental Asia which has invaded and established in Hawaii and Tanzania and has been recovered in detection trapping in California. It is largely non-responsive to the male

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service