Skip to Content
Merck
All Photos(1)

Key Documents

00300590

Rutin trihydrate

primary reference standard

Synonym(s):

Quercetin-3-rutinoside trihydrate, Vitamin P trihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€235.00

€235.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€235.00

About This Item

Empirical Formula (Hill Notation):
C27H30O16 · 3H2O
CAS Number:
Molecular Weight:
664.56
Beilstein:
75455
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€235.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

SMILES string

O.O.O.C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(Oc4cc(O)cc(O)c4C3=O)c5ccc(O)c(O)c5)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H30O16.3H2O/c1-8-17(32)20(35)22(37)26(40-8)39-7-15-18(33)21(36)23(38)27(42-15)43-25-19(34)16-13(31)5-10(28)6-14(16)41-24(25)9-2-3-11(29)12(30)4-9;;;/h2-6,8,15,17-18,20-23,26-33,35-38H,7H2,1H3;3*1H2/t8-,15+,17-,18+,20+,21-,22+,23+,26+,27-;;;/m0.../s1

InChI key

NLLBWFFSGHKUSY-JPRRWYCFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference standard in the analysis of herbal medicinal products.

Other Notes

This compound is commonly found in plants of the genus: achillea crataegus echinacea eucalyptus ginkgo glycyrrhiza hedera humulus hypericum lycium pimpinella primula sambucus serenoa

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nattapon Supkamonseni et al.
Indian journal of experimental biology, 52(10), 965-971 (2014-10-28)
In vitro study revealed that pancreatic lipase inhibitory activity of C. asiatica extract was significantly higher than rutin but lower than orlistat, an anti-obesity drug. alpha-Amylase inhibitory activities of C. asiatica extract and rutin were significantly lower than acarbose, an
Snezana Agatonovic-Kustrin et al.
Journal of chromatography. A, 1385, 103-110 (2015-02-11)
The present study describes a simple high performance thin layer chromatographic (HPTLC) method for the simultaneous quantification of apigenin, chamazulene, bisabolol and the use of DPPH free radical as a post-chromatographic derivatization agent to compare the free radical scavenging activities

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service