Skip to Content
Merck
All Photos(1)

Key Documents

40-0102

Sigma-Aldrich

pH standard solution

pH 4.01 (25 °C)

Synonym(s):

pH 4.01 buffer solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700

availability

available only in Japan

pH

4.01 (25 °C)

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service