37-0190
Zinc acetate dihydrate
JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Zn(CH3COO)2 · 2H2O
CAS Number:
Molecular Weight:
219.51
Beilstein:
3732513
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
JIS special grade
form:
solid
Recommended Products
grade
JIS special grade
Assay
≥99.0%
form
solid
reaction suitability
reagent type: catalyst
core: zinc
availability
available only in Japan
storage temp.
15-25°C
SMILES string
O.O.CC(=O)O[Zn]OC(C)=O
InChI
1S/2C2H4O2.2H2O.Zn/c2*1-2(3)4;;;/h2*1H3,(H,3,4);2*1H2;/q;;;;+2/p-2
InChI key
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Robert Krysiak et al.
The Yale journal of biology and medicine, 85(2), 249-254 (2012-06-28)
Wilson's disease is a rare genetic disorder of copper metabolism. The difference in copper tissue accumulation is responsible for the various clinical manifestations of this disorder. If left untreated, Wilson's disease progresses to hepatic failure, severe neurological disability, and even
Pier Francesco Porciani et al.
The Journal of clinical dentistry, 23(3), 76-79 (2012-12-06)
A controlled, clinical, double-blind study was conducted to assess the efficacy of a sugar-free chewing gum containing zinc acetate and magnolia bark extract (MBE) on oral volatile sulfur-containing compounds (VSC) versus a placebo sugar-free chewing gum for two hours. To
Alim Solmaz et al.
Langmuir : the ACS journal of surfaces and colloids, 28(28), 10592-10596 (2012-06-15)
Tapping mode atomic force microscopy (TM-AFM) enables mapping of chemical composition at the nanoscale by taking advantage of the variation in phase angle shift arising from an embedded second phase. We demonstrate that phase contrast can be attributed to the
L Ruby Nirmala et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 97, 673-677 (2012-08-08)
Single crystals of pure bis thiourea zinc acetate (BTZA) and lithium chloride (LiCl) doped BTZA have been synthesized and grown successfully from aqueous solutions by slow evaporation technique. The single crystals thus grown were characterized by different techniques such as
R Dhariwal et al.
Journal of the Indian Society of Pedodontics and Preventive Dentistry, 30(1), 85-88 (2012-05-09)
Oral submucous fibrosis is a chronic debilitating disorder which is usually seen in adults with areca nut chewing habit. A rapid upsurge in the popularity of commercial areca nut products, especially among the younger generation, is a cause of grave
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service