Skip to Content
Merck
All Photos(1)

Key Documents

34-0015

Sigma-Aldrich

Wijs (iodine monochloride) solution | Chloroiodide | 7790-99-0 solution

Synonym(s):

Iodine monochloride solution, Chloroiodide solution, Wijs solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICl
CAS Number:
Molecular Weight:
162.36
Beilstein:
3587194
MDL number:
UNSPSC Code:
12352307
PubChem Substance ID:
grade:
SAJ special grade
form:
liquid
Pricing and availability is not currently available.

grade

SAJ special grade

form

liquid

availability

available only in Japan

storage temp.

15-25°C

SMILES string

ClI

InChI

1S/ClI/c1-2

InChI key

QZRGKCOWNLSUDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

for the iodine value determination

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

102.2 °F

Flash Point(C)

39 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Suman Lata et al.
Analytical biochemistry, 313(1), 155-159 (2003-02-11)
A single-step procedure was developed for the incorporation of iodoazide into oleic acid, triolein, and phosphatidylcholine. Iodoazide was generated using [125I]iodomonochloride and sodium azide that was found to add stereospecifically in a variety of olefins. Photoreactive and radiolabeled triacylglycerol and
Cell surface labeling by iodine monochloride.
E J Victoria et al.
The International journal of biochemistry, 13(9), 1011-1017 (1981-01-01)
C P Coyne et al.
American journal of veterinary research, 46(12), 2578-2581 (1985-12-01)
Two methods were analyzed for the rapid extraction of equine fibrinogen from fresh plasma, using ammonium sulfate-sodium phosphate buffer. Fibrinogen from each of these 2 methods was then radiolabeled with 125I (half-life = 60.2 days, gamma = 35 keV), using
Iodine monochloride (IC1) iodination techniques.
M A Contreras et al.
Methods in enzymology, 92, 277-292 (1983-01-01)
L C Knight et al.
Thrombosis and haemostasis, 46(3), 593-596 (1981-10-01)
The properties of human fibrinogen labeled with 125-Iodine using Iodogen (1, 3, 4, 6-tetrachloro-3 alpha, 6 alpha-diphenylglycoluril) as an oxidizing agent were compared with those of an iodine monochloride labeled counterpart. It was found that thrombin clottability, binding to staphylococci

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service