Skip to Content
Merck
All Photos(1)

Key Documents

32-0140

Sigma-Aldrich

Fluorescein sodium salt

SAJ first grade

Synonym(s):

Acid Yellow 73, D&C;Yellow No. 8, NaFl, NaFluo, Sodium fluorescein, Uranine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H10Na2O5
CAS Number:
Molecular Weight:
376.27
Colour Index Number:
45350
Beilstein:
3833041
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

form

solid

availability

available only in Japan

pH

~8.3 (20 °C, 10 g/L)

fluorescence

λex 460 nm; λem 515 nm(lit.)

storage temp.

15-25°C

SMILES string

[Na+].[Na+].[O-]c1ccc2c(Oc3cc([O-])ccc3C24OC(=O)c5ccccc45)c1

InChI

1S/C20H12O5.2Na/c21-11-5-7-15-17(9-11)24-18-10-12(22)6-8-16(18)20(15)14-4-2-1-3-13(14)19(23)25-20;;/h1-10,21-22H;;/q;2*+1/p-2

InChI key

RGPLGPBQJOQFJS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

423.7 °F - Pensky-Martens closed cup

Flash Point(C)

217.6 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher Price et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 26(2), 277-285 (2010-08-18)
Since proposed by Piekarski and Munro in 1977, load-induced fluid flow through the bone lacunar-canalicular system (LCS) has been accepted as critical for bone metabolism, mechanotransduction, and adaptation. However, direct unequivocal observation and quantification of load-induced fluid and solute convection
Sara Seabrooke et al.
American journal of physiology. Cell physiology, 305(5), C558-C567 (2013-06-28)
The blood-brain barrier (BBB) physiologically isolates the brain from the blood and, thus, plays a vital role in brain homeostasis. Ion transporters play a critical role in this process by effectively regulating access of chemicals to the brain. Organic anion-transporting
Tomoaki Murakami et al.
Ophthalmology, 120(12), 2596-2603 (2013-08-21)
To study the association between the fluorescence levels on fluorescein angiography images and the characteristics on spectral-domain optical coherence tomography (SD OCT) images in diabetic macular edema (DME). Retrospective, observational, cross-sectional study. One hundred sixty-seven consecutive eyes of 116 patients
Wee Yang Ng et al.
Biomicrofluidics, 3(2), 22405-22405 (2009-08-21)
This paper investigates the phenomenon of Faradaic charging in ac electrokinetics. Faradaic reactions were suggested as a key effect responsible for the reversal of pumping direction in ac micropumps. However, this hypothesis has yet to be proven convincingly and directly.
Beau J Fenner et al.
Investigative ophthalmology & visual science, 54(13), 8013-8019 (2013-11-14)
To determine the reliability of single- and double-zone corneal fluorescent staining compared with five-zone analysis for the prediction of dry eye disease. Prospective study of 510 subjects with dry eye disease characterized using corneal fluorescein staining, Schirmer scores, and tear

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service